missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-piperidone, 98+%

Manufacturer:  Alfa Aesar™ A1239022

 View more versions of this product

Catalog No. AAA1239022



Description

Description

Applications
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.

Solubility
Soluble in water (12 g/100ml at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

1-Benzyl-4-piperidone
248°C
3612-20-2
MFCD00006192
1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
C1CN(CCC1=O)CC2=CC=CC=C2
189.258
189.26
>110°C (230°F)
100g
C12H15NO
128556
SJZKULRDWHPHGG-UHFFFAOYSA-N
1-benzylpiperidin-4-one
19220
≥98%

Chemical Identifiers

3612-20-2
189.258
SJZKULRDWHPHGG-UHFFFAOYSA-N
19220
C1CN(CCC1=O)CC2=CC=CC=C2
C12H15NO
MFCD00006192
1-benzyl-4-piperidone, n-benzyl-4-piperidone, 4-piperidinone, 1-phenylmethyl, 1-benzyl-4-piperidinone, 1-benzylpiperidone, 1-benzyl-4-oxopiperidine, 1-benzyl-piperidin-4-one, n-benzyl-4-piperidine, n-benzyl-4-piperidinone, n-benzylpiperidin-4-one
1-benzylpiperidin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2°C to 8°C

SDS Documents