Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-methylpiperazine hydrochloride monohydrate, 98%

$72.80 - $241.66

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-methylpiperazine hydrochloride monohydrate
CAS 374898-00-7
Molecular Formula C12H18N2·HCl·H2O
Formula Weight 244.76 (226.75 Anhydrous)
MDL Number MFCD03093089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1911109

  SDS
alfa aesar™
L1911109
10g Each for $72.80

AAL1911118

  SDS
alfa aesar™
L1911118
50g Each for $241.66
Description & Specifications

Specifications

Formula Weight 244.76 (226.75 Anhydrous)
Chemical Name or Material 1-Benzyl-4-methylpiperazine hydrochloride monohydrate
Molecular Formula C12H18N2·HCl·H2O
MDL Number MFCD03093089
CAS 374898-00-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Melting Point 111° to 114°C
Quantity 10g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.