Please login to your online account to display your discounted pricing

1-Benzyl-4-hydroxypiperidine, 97%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  166831000

 View more versions of this product

Catalog No. AC166831000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 191.27
Quantity 100g
Color White to yellow
EINECS Number 225-226-9
TSCA TSCA
Melting Point 60° to 65°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H17NO
MDL Number MFCD00006503
CAS 4727-72-4
Packaging Glass bottle
Synonym 1-Benzyl-4-piperidinol
Infrared Spectrum Authentic
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Name Note 97%
CAS 4727-72-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00006503
Synonym 1-Benzyl-4-piperidinol
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents