missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-4-hydroxypiperidine, 97%, ACROS Organics™

Manufacturer:  Acros Organics 166830250

 View more versions of this product

Catalog No. AC166830250



Specifications

Specifications

1-Benzyl-4-hydroxypiperidine
Glass bottle
60°C to 65°C
97%
96.0
97%
MFCD00006503
BPPZXJZYCOETDA-UHFFFAOYSA-N
1-benzylpiperidin-4-ol
78461
Crystalline Powder
Authentic
White to Yellow
25g
4727-72-4
100.0
C12H17NO
1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
C1CN(CCC1O)CC2=CC=CC=C2
191.274
191.27
≥96.0%

Chemical Identifiers

4727-72-4
191.274
BPPZXJZYCOETDA-UHFFFAOYSA-N
78461
C1CN(CCC1O)CC2=CC=CC=C2
C12H17NO
MFCD00006503
1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
1-benzylpiperidin-4-ol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 225-226-9

TSCA : TSCA

SDS Documents