Please login to your online account to display your discounted pricing

1-Benzyl-4-hydroxypiperidine, 97%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC166830050

 View more versions of this product

Catalog No. AC166830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.27
Assay ≥96.0%
Physical Form Crystalline Powder
Quantity 5g
Color White to yellow
EINECS Number 225-226-9
Melting Point 60° to 65°C
TSCA TSCA
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 4727-72-4
Molecular Formula C12H17NO
Assay Percent Range 97%
MDL Number MFCD00006503
Name Note 97%
Packaging Glass bottle
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Infrared Spectrum Authentic
Synonym 1-Benzyl-4-piperidinol
Health Hazard 1 Warning

Specifications

Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Name Note 97%
CAS 4727-72-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00006503
Synonym 1-Benzyl-4-piperidinol
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents