Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-hydroxypiperidine, 97%

$164.24 - $527.36

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
CAS 4727-72-4
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00006503
Beilstein 146118
Synonym 1-Benzyl-4-piperidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1093214

  SDS
alfa aesar™
A1093214
25g Each for $164.24

AAA1093222

  SDS
alfa aesar™
A1093222
100g Each for $527.36
Description & Specifications

Specifications

Formula Weight 191.27
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00006503
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-Benzyl-4-piperidinol
Boiling Point 121° to 123°C (0.7mmHg)
Health Hazard 1 Warning
Chemical Name or Material 1-Benzyl-4-hydroxypiperidine
Molecular Formula C12H17NO
Assay 97%
CAS 4727-72-4
Melting Point 60° to 64°C
Beilstein 146118
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.