Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, tech. 90%

$125.51 - $125.51

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride
CAS 52763-21-0
Purity Grade Technical Grade
Molecular Formula C15H19NO3·HCl
Formula Weight 297.78
MDL Number MFCD00012792
Beilstein 3749159
Synonym 1-Benzyl-4-carboethoxy-3-piperidone hydrochloride; Ethyl 1-benzyl-3-piperidone-4-carboxylate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0747206

  SDS
alfa aesar™
L0747206
5g Each for $125.51
Description & Specifications

Specifications

Formula Weight 297.78
Chemical Name or Material 1-Benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride
Beilstein 3749159
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 52763-21-0
Health Hazard 1 Warning
MDL Number MFCD00012792
Assay 90%
Molecular Formula C15H19NO3·HCl
Purity Grade Technical Grade
Synonym 1-Benzyl-4-carboethoxy-3-piperidone hydrochloride; Ethyl 1-benzyl-3-piperidone-4-carboxylate hydrochloride
Melting Point ≈158°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.