Please login to your online account to display your discounted pricing

1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride, 99%, ACROS Organics™

$121.63 - $121.63

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
CAS 71258-18-9
CAS Min % 98.5
CAS Max % 100.0
Assay 99.0 to 101.0%
Assay Percent Range 99%
Molecular Formula C19H20N2·HCl
Formula Weight 312.83
MDL Number MFCD00035361
Synonym 4-Phenyl-1-(phenylmethyl)-4-piperidinecarbonitrile
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC135860250

  SDS
thermo fisher scientific
135860250
Glass bottle 25g Each for $121.63
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 312.83
Quantity 25g
Color White to beige
Assay 99.0 to 101.0%
CAS Max % 100.0
Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
Assay Percent Range 99%
Molecular Formula C19H20N2·HCl
CAS 71258-18-9
Synonym 4-Phenyl-1-(phenylmethyl)-4-piperidinecarbonitrile
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00035361
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.