Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride, 99%

$58.81 - $409.45

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
CAS 56243-25-5
Molecular Formula C19H20N2·HCl
Formula Weight 312.85
MDL Number MFCD00035361
Synonym 1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741106

  SDS
alfa aesar™
H2741106
5g Each for $58.81

AAH2741114

  SDS
alfa aesar™
H2741114
25g Each for $163.16

AAH2741122

  SDS
alfa aesar™
H2741122
100g Each for $409.45
Description & Specifications

Specifications

Formula Weight 312.85
Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine hydrochloride
Assay 99%
Molecular Formula C19H20N2·HCl
MDL Number MFCD00035361
CAS 56243-25-5
Synonym 1-Benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride
Melting Point 261° to 264°C
Health Hazard 1 Danger
Quantity 5g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.