missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzyl-4-cyano-4-hydroxypiperidine, 98%, ACROS Organics™
Chemical Identifiers
CAS | 6094-60-6 |
---|---|
Molecular Formula | C13H16N2O |
Molecular Weight (g/mol) | 216.28 |
InChI Key | XQRLXUYZKZXSBN-UHFFFAOYSA-N |
Synonym | 1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-cyano-4-hydroxypiperidine, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile, acmc-20aoxx, maybridge1_006722 |
PubChem CID | 80189 |
IUPAC Name | 1-benzyl-4-hydroxypiperidine-4-carbonitrile |
SMILES | C1CN(CCC1(C#N)O)CC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC136860100
|
Thermo Fisher Scientific
136860100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
6094-60-6 | |
216.28 | |
1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-cyano-4-hydroxypiperidine, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile, acmc-20aoxx, maybridge1_006722 | |
1-benzyl-4-hydroxypiperidine-4-carbonitrile |
C13H16N2O | |
XQRLXUYZKZXSBN-UHFFFAOYSA-N | |
80189 | |
C1CN(CCC1(C#N)O)CC2=CC=CC=C2 |
Specifications
Authentic | |
Yellow to Beige | |
10g | |
97.5 | |
97.5% min. (GC) | |
1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-cyano-4-hydroxypiperidine, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile, acmc-20aoxx, maybridge1_006722 | |
C1CN(CCC1(C#N)O)CC2=CC=CC=C2 | |
216.28 | |
216.28 | |
98% |
Glass bottle | |
95°C to 102°C | |
6094-60-6 | |
100.0 | |
C13H16N2O | |
XQRLXUYZKZXSBN-UHFFFAOYSA-N | |
1-benzyl-4-hydroxypiperidine-4-carbonitrile | |
80189 | |
Crystalline Powder and Chunks | |
1-Benzyl-4-cyano-4-hydroxypiperidine, 98% |
Safety and Handling
EINECSNumber : 228-043-2
TSCA : TSCA