Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-4-chlorophthalazine, 99%

$127.18 - $439.97

Chemical Identifiers

Chemical Name or Material 1-Benzyl-4-chlorophthalazine
CAS 40848-53-1
Molecular Formula C15H11ClN2
Formula Weight 254.72
MDL Number MFCD00172387
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5049403

  SDS
alfa aesar™
H5049403
1g Each for $439.97

AAH50494MD

  SDS
alfa aesar™
H50494MD
250mg Each for $127.18
Description & Specifications

Specifications

Formula Weight 254.72
Chemical Name or Material 1-Benzyl-4-chlorophthalazine
Assay 99%
Quantity 1g
CAS 40848-53-1
MDL Number MFCD00172387
Molecular Formula C15H11ClN2

Applications
1-Benzyl-4-chlorophthalazine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.