Please login to your online account to display your discounted pricing

1-Benzyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Benzylpyrazole-4-boronic acid pinacol ester
CAS 761446-45-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C16H21BN2O2
Formula Weight 284.16
MDL Number MFCD03789252
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC371760010

  SDS
thermo fisher scientific
AC371760010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC371762500

  SDS
thermo fisher scientific
AC371762500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.16
Physical Form Crystals or Crystalline Powder
Assay ≥96.0%
Quantity 1g
Color Off-white to light yellow
Melting Point 86° to 91°C
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-Benzylpyrazole-4-boronic acid pinacol ester
Molecular Formula C16H21BN2O2
Assay Percent Range 97%
CAS 761446-45-1
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD03789252
Packaging Glass bottle
Infrared Spectrum Authentic