Please login to your online account to display your discounted pricing

1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid, ≥95%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid
CAS 100957-85-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C15H18N2O2
Formula Weight 258.32
MDL Number MFCD02090479
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB03031EA

 
fisher scientific company
SPB03031EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 258.32
Quantity 10g
Assay 95%
CAS Min % 95.0
Chemical Name or Material 1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carboxylic acid
Assay Percent Range 95%
Molecular Formula C15H18N2O2
MDL Number MFCD02090479
CAS 100957-85-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Amber Glass Bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.