Please login to your online account to display your discounted pricing

1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carbonyl chloride, 95%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB03039CB

 View more versions of this product

Catalog No. SPB03039CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 276.77
Quantity 250mg
Assay 95%
CAS Min % 95.0
CAS 160233-29-4
Chemical Name or Material 1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carbonyl chloride
MDL Number MFCD00084892
Packaging Amber Glass Bottle
Assay Percent Range 95%
Molecular Formula C15H17ClN2O
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Benzyl-3-(tert-butyl)-1H-pyrazole-5-carbonyl chloride
CAS 160233-29-4
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C15H17ClN2O
Formula Weight 276.77
MDL Number MFCD00084892
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents