Learn More
1-Benzyl-3-pyrroline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1949603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Benzyl-3-pyrroline | |
0.837 | |
92°C (197°F) | |
1.542 | |
1 g | |
111101 | |
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline | |
C1C=CCN1CC2=CC=CC=C2 | |
159.232 | |
159.23 |
6913-92-4 | |
60°C (1 mmHg) | |
C11H13N | |
MFCD00012324 | |
UN2735 | |
Air sensitive | |
LRFHKHHUKGZIGE-UHFFFAOYSA-N | |
1-benzyl-2,5-dihydropyrrole | |
561506 | |
≥98% |
Chemical Identifiers
6913-92-4 | |
159.232 | |
LRFHKHHUKGZIGE-UHFFFAOYSA-N | |
561506 | |
C1C=CCN1CC2=CC=CC=C2 |
C11H13N | |
MFCD00012324 | |
1-benzyl-3-pyrroline, 1-benzyl-2,5-dihydro-1h-pyrrole, n-benzylpyrroline, n-benzyl-3-pyrroline, 1h-pyrrole, 2,5-dihydro-1-phenylmethyl, maleimide-related compound 16, l-benzyl-3-pyrroline, n-benzyl 3-pyrroline, 1-benzylpyrroline | |
1-benzyl-2,5-dihydropyrrole |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only