Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-pyrrolidinone, 98%

$1,089.39 - $1,089.39

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-pyrrolidinone
CAS 775-16-6
Molecular Formula C11H13NO
Formula Weight 175.23
MDL Number MFCD00005342
Synonym N-Benzyl-3-pyrrolidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1342614

  SDS
alfa aesar™
A1342614
25g Each for $1,089.39
Description & Specifications

Specifications

Formula Weight 175.23
Assay 98%
Quantity 25g
Chemical Name or Material 1-Benzyl-3-pyrrolidinone
Boiling Point 77° to 78°C (0.01mmHg)
Flash Point >110°C (230°F)
CAS 775-16-6
Recommended Storage 2° to 8°C
Molecular Formula C11H13NO
MDL Number MFCD00005342
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Synonym N-Benzyl-3-pyrrolidone
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C