Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-piperidone hydrochloride hydrate

$57.73 - $720.38

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-piperidone hydrochloride hydrate
CAS 50606-58-1
Molecular Formula C12H15NO·HCl·xH2O
Formula Weight 225.72 (Anhydrous)
MDL Number MFCD00150096
Beilstein 3705146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1284903

  SDS
alfa aesar™
A1284903
1g Each for $57.73

AAA1284914

  SDS
alfa aesar™
A1284914
25g Each for $720.38
Description & Specifications

Specifications

Formula Weight 225.72 (Anhydrous)
Chemical Name or Material 1-Benzyl-3-piperidone hydrochloride hydrate
Molecular Formula C12H15NO·HCl·xH2O
MDL Number MFCD00150096
CAS 50606-58-1
Beilstein 3705146
Melting Point 105° to 107°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C