missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-3-piperidone hydrate hydrochloride, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC166920010

 View more versions of this product

Catalog No. AC166920010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Benzyl-3-piperidone hydrate hydrochloride
Glass bottle
Brown to Cream
95%
94.0
95%
MFCD00150096
OVWSFXNSJDMRPV-UHFFFAOYSA-N
1-benzylpiperidin-3-one;hydrochloride
3084924
Powder
Authentic
8% max. (K.F.)
1g
50606-58-1
100.0
C12H16ClNO
1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl
225.716
225.72
≥94% (on free base)

Chemical Identifiers

50606-58-1
225.716
OVWSFXNSJDMRPV-UHFFFAOYSA-N
3084924
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl
C12H16ClNO
MFCD00150096
1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726
1-benzylpiperidin-3-one;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 256-646-0

TSCA : TSCA

SDS Documents