Learn More
1-Benzyl-3-piperidone hydrate hydrochloride, 95%, ACROS Organics™
$634.02 - $634.02
Chemical Identifiers
CAS | 50606-58-1 |
---|---|
Molecular Formula | HCl·xH2O |
Molecular Weight (g/mol) | 225.72 |
MDL Number | MFCD00150096 |
InChI Key | OVWSFXNSJDMRPV-UHFFFAOYSA-N |
Synonym | 1-benzyl-3-piperidinone hydrate hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hcl hydrate, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzylpiperidin-3-one hydrochloride, 3-piperidinone, 1-phenylmethyl-, hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726 |
PubChem CID | 3084924 |
IUPAC Name | 1-benzylpiperidin-3-one;hydrochloride |
SMILES | C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC166920010
|
Thermo Fisher Scientific
AC166920010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC166920250
|
Thermo Fisher Scientific
166920250 |
25g | Glass bottle |
Each for $634.02
|
|
Chemical Identifiers
50606-58-1 | |
225.72 | |
OVWSFXNSJDMRPV-UHFFFAOYSA-N | |
3084924 | |
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl |
HCl·xH2O | |
MFCD00150096 | |
1-benzyl-3-piperidinone hydrate hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hcl hydrate, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzylpiperidin-3-one hydrochloride, 3-piperidinone, 1-phenylmethyl-, hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726 | |
1-benzylpiperidin-3-one;hydrochloride |
Specifications
Authentic | |
8% max. (K.F.) | |
170°C to 176°C | |
50606-58-1 | |
100.0 | |
HCl·xH2O | |
1-benzyl-3-piperidinone hydrate hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hcl hydrate, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzylpiperidin-3-one hydrochloride, 3-piperidinone, 1-phenylmethyl-, hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726 | |
OVWSFXNSJDMRPV-UHFFFAOYSA-N | |
1-benzylpiperidin-3-one;hydrochloride | |
3084924 | |
Powder | |
1-Benzyl-3-piperidone hydrate hydrochloride, 95% |
Glass bottle | |
Brown to Cream | |
1g | |
94.0 | |
94% min. (on free base) (GC) | |
MFCD00150096 | |
Solubility in water: soluble | |
C1CC(=O)CN(C1)CC2=CC=CC=C2.Cl | |
225.72 | |
225.72 | |
95% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 256-646-0
TSCA : TSCA