Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-methylpiperazine hydrochloride, 98+%

$38.86 - $430.93

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-methylpiperazine hydrochloride
Molecular Formula C12H18N2·HCl
Formula Weight 226.75
MDL Number MFCD09842143
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3016903

  SDS
alfa aesar™
H3016903
1g Each for $38.86

AAH3016906

  SDS
alfa aesar™
H3016906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3016914

  SDS
alfa aesar™
H3016914
25g Each for $430.93
Description & Specifications

Specifications

Formula Weight 226.75
Health Hazard 1 Warning
Chemical Name or Material 1-Benzyl-3-methylpiperazine hydrochloride
Assay ≥98%
Molecular Formula C12H18N2·HCl
Quantity 1g
MDL Number MFCD09842143
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.