missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-methylimidazolium hexafluorophosphate, 97%

Manufacturer:  Alfa Aesar™ H5954806

 View more versions of this product

Catalog No. AAH5954806



Specifications

Specifications

1-Benzyl-3-methylimidazolium hexafluorophosphate
White
433337-11-2
MFCD07784448
VVCQOUQQPZPIKL-UHFFFAOYSA-N
1-benzyl-3-methylimidazol-3-ium;hexafluorophosphate
11461285
Solid
Moisture Sensitive
5g
C11H13F6N2P
1-benzyl-3-methylimidazolium hexafluorophosphate, 1-benzyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 3-benzyl-1-methylimidazol-1-ium hexafluorophosphate, ksc943e0j, 1-benzyl-3-methylimid-azolium hexafluorophosphate, 1-methyl-3-benzylimidazolium hexafluorophosphate, 1-benzyl-3-methylimidazol-1-ium hexafluorophosphate, 1-benzyl-3-methylimidazolium hexafluorophosphate, 1h-imidazolium, 1-methyl-3-phenylmethyl-, hexafluorophosphate 1-
C[N+]1=CN(C=C1)CC2=CC=CC=C2.F[P-](F)(F)(F)(F)F
318.203
318.2
97%

Chemical Identifiers

433337-11-2
318.203
VVCQOUQQPZPIKL-UHFFFAOYSA-N
11461285
C[N+]1=CN(C=C1)CC2=CC=CC=C2.F[P-](F)(F)(F)(F)F
C11H13F6N2P
MFCD07784448
1-benzyl-3-methylimidazolium hexafluorophosphate, 1-benzyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 3-benzyl-1-methylimidazol-1-ium hexafluorophosphate, ksc943e0j, 1-benzyl-3-methylimid-azolium hexafluorophosphate, 1-methyl-3-benzylimidazolium hexafluorophosphate, 1-benzyl-3-methylimidazol-1-ium hexafluorophosphate, 1-benzyl-3-methylimidazolium hexafluorophosphate, 1h-imidazolium, 1-methyl-3-phenylmethyl-, hexafluorophosphate 1-
1-benzyl-3-methylimidazol-3-ium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents