Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-methylimidazolium hexafluorophosphate, 97%

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-methylimidazolium hexafluorophosphate
CAS 433337-11-2
Formula Weight 318.2
MDL Number MFCD07784448
Synonym 1-Methyl-3-benzylimidazolium hexafluorophosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5954806

  SDS
alfa aesar™
H5954806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5954818

  SDS
alfa aesar™
H5954818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 318.2
CAS 433337-11-2
Health Hazard 1 Warning
Synonym 1-Methyl-3-benzylimidazolium hexafluorophosphate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Sensitivity Moisture Sensitive
MDL Number MFCD07784448
Quantity 5g
Physical Form Solid
Color White
Chemical Name or Material 1-Benzyl-3-methylimidazolium hexafluorophosphate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.