missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-3-methyl-4-piperidone, TCI America™

$88.61 - $261.25

Chemical Identifiers

CAS 34737-89-8
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD00044806
InChI Key OVQAJYCAXPHYNV-UHFFFAOYSA-N
Synonym 1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
PubChem CID 560965
IUPAC Name 1-benzyl-3-methylpiperidin-4-one
SMILES CC1CN(CCC1=O)CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B44651G View Documents TCI America
B44651G
1g Each for $88.61
 
B44655G View Documents TCI America
B44655G
5g Each for $261.25
 
Specifications

Chemical Identifiers

34737-89-8
203.285
OVQAJYCAXPHYNV-UHFFFAOYSA-N
560965
CC1CN(CCC1=O)CC2=CC=CC=C2
C13H17NO
MFCD00044806
1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
1-benzyl-3-methylpiperidin-4-one

Specifications

99°C
1g
C13H17NO
1760
OVQAJYCAXPHYNV-UHFFFAOYSA-N
1-benzyl-3-methylpiperidin-4-one
560965
Clear Liquid at 20°C
1-Benzyl-3-methyl-4-piperidone
Yellow
34737-89-8
MFCD00044806
1-benzyl-3-methyl-4-piperidone, 4-piperidinone, 3-methyl-1-phenylmethyl, 1-benzyl-3-methyl-4-piperidinone, 4-piperidone, 1-benzyl-3-methyl, n-benzyl-3-methyl-4-piperidone, 3-methyl-1-phenylmethyl-4-piperidinone, 3-methyl-1-benzylpiperidin-4-one, ksc506m4r, 1-benzyl-3-methyl-piperidin-4one, 1-benzyl-3-methyl-piperidin4-one
CC1CN(CCC1=O)CC2=CC=CC=C2
203.285
203.29
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator