Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-iodo-7-azaindole, 97%

$212.46 - $212.46

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-iodo-7-azaindole
CAS 1093759-51-3
Molecular Formula C14H11IN2
Formula Weight 334.16
MDL Number MFCD09834873
Beilstein 24597035
Synonym 1-Benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5167506

  SDS
alfa aesar™
H5167506
5g Each for $212.46
Description & Specifications

Specifications

Formula Weight 334.16
Quantity 5g
Molecular Formula C14H11IN2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD09834873
Synonym 1-Benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
CAS 1093759-51-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Benzyl-3-iodo-7-azaindole
Assay 97%
Health Hazard 1 Warning
Beilstein 24597035
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.