Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-hydroxypiperidine, 99%

$64.93 - $248.62

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-hydroxypiperidine
CAS 14813-01-5
Molecular Formula C12H17NO
Formula Weight 191.27
MDL Number MFCD00023734
Beilstein 135964
Synonym 1-Benzyl-3-piperidinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1637503

  SDS
alfa aesar™
L1637503
1g Each for $64.93

AAL1637506

  SDS
alfa aesar™
L1637506
5g Each for $248.62
Description & Specifications

Specifications

Formula Weight 191.27
Chemical Name or Material 1-Benzyl-3-hydroxypiperidine
Molecular Formula C12H17NO
MDL Number MFCD00023734
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 99%
Beilstein 135964
Boiling Point 140° to 142°C (6mmHg)
Health Hazard 1 Warning
Quantity 1g
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
CAS 14813-01-5
Synonym 1-Benzyl-3-piperidinol
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.