Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-hydroxy-1H-indazole, 98%

$92.53 - $250.12

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-hydroxy-1H-indazole
CAS 2215-63-6
Molecular Formula C14H12N2O
Formula Weight 224.26
MDL Number MFCD01631173
Synonym 1-Benzyl-1,2-dihydro-3H-indazol-3-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2536614

  SDS
alfa aesar™
B2536614
25g Each for $92.53

AAB2536622

  SDS
alfa aesar™
B2536622
100g Each for $250.12
Description & Specifications

Specifications

Formula Weight 224.26
CAS 2215-63-6
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-Benzyl-3-hydroxy-1H-indazole
Synonym 1-Benzyl-1,2-dihydro-3H-indazol-3-one
Quantity 25g
Assay 98%
Melting Point 164° to 166°C
MDL Number MFCD01631173
Molecular Formula C14H12N2O
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.