Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride hydrate, 97%, ca 10% water

$67.11 - $989.76

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride hydrate
CAS 1454-53-1
Molecular Formula C15H19NO3·HCl·xH2O
Formula Weight 297.78 (Anhydrous)
MDL Number MFCD00012800
Beilstein 3918980
Synonym 1-Benzyl-3-carboethoxy-4-piperidone hydrochloride hydrate; Ethyl 1-benzyl-4-piperidone-3-carboxylate hydrochloride hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1354209

  SDS
alfa aesar™
A1354209
10g Each for $67.11

AAA1354230

  SDS
alfa aesar™
A1354230
250g Each for $989.76
Description & Specifications

Specifications

Formula Weight 297.78 (Anhydrous)
Chemical Name or Material 1-Benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride hydrate
Assay 97%
Assay Percent Notes ≈10% water
Molecular Formula C15H19NO3·HCl·xH2O
MDL Number MFCD00012800
CAS 1454-53-1
Beilstein 3918980
Synonym 1-Benzyl-3-carboethoxy-4-piperidone hydrochloride hydrate; Ethyl 1-benzyl-4-piperidone-3-carboxylate hydrochloride hydrate
Melting Point ≈170°C (dec.)
Health Hazard 1 Warning
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.