Learn More
1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97%, ACROS Organics™
$114.67 - $114.67
Chemical Identifiers
CAS | 1454-53-1 |
---|---|
Molecular Formula | C15H19NO3·HCl |
Molecular Weight (g/mol) | 297.77 |
MDL Number | MFCD00012800 |
InChI Key | YPFMNHZRNXPYBG-UHFFFAOYSA-N |
Synonym | 1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride |
PubChem CID | 102623 |
IUPAC Name | ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride |
SMILES | CCOC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC105900250
|
Thermo Fisher Scientific
105900250 |
25g | Glass bottle |
Each for $114.67
|
|
AC105901000
|
Thermo Fisher Scientific
105901000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1454-53-1 | |
297.77 | |
YPFMNHZRNXPYBG-UHFFFAOYSA-N | |
102623 | |
CCOC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl |
C15H19NO3·HCl | |
MFCD00012800 | |
1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride | |
ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride |
Specifications
Authentic | |
12% max. (K.F.) | |
160.0°C to 170.0°C | |
1454-53-1 | |
100.0 | |
C15H19NO3·HCl | |
22, II, 216 | |
YPFMNHZRNXPYBG-UHFFFAOYSA-N | |
ethyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride | |
102623 | |
Powder | |
1-Benzyl-3-carbethoxy-4-piperidone hydrochloride, 97% |
Glass bottle | |
Cream to Pink | |
25g | |
96.0 | |
96% min. (on dry substance) (Argentometry) | |
MFCD00012800 | |
1-benzyl-3-carbethoxy-4-piperidone hcl, 1-benzyl-3-carbethoxy-4-piperidone hydrochloride, 1-benzyl-3-carbethoxy-piperidone hcl, 1-benzyl-3-carboethoxy-4-piperidone hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-oxopiperidine hydrochloride, 1-benzyl-3-ethoxycarbonyl-4-piperidone hcl, 1-benzyl-3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, ethyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride | |
CCOC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl | |
297.77 | |
297.77 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 215-929-9
TSCA : TSCA