Please login to your online account to display your discounted pricing

1-Benzyl-3-bromo-5-fluoro-1H-1,2,4-triazole 97+%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3-bromo-5-fluoro-1H-1, 2, 4-triazole
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H7BrFN3
Formula Weight 256.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07869CB

 
fisher scientific company
MO07869CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 256.08
Quantity 250mg
Assay 97%
Chemical Name or Material 1-Benzyl-3-bromo-5-fluoro-1H-1, 2, 4-triazole
Assay Percent Range 97%
Molecular Formula C9H7BrFN3
CAS 214540-43-9
Packaging Amber Glass Bottle