Please login to your online account to display your discounted pricing

1-benzyl-3,5-dibromo-1H-1,2,4-triazole, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7Br2N3
Formula Weight 316.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07858DA

  SDS
fisher scientific company
MO07858DA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 316.98
Quantity 1g
Assay 95%
Chemical Name or Material 1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Assay Percent Range 95%
Molecular Formula C9H7Br2N3
CAS 106724-85-0
Health Hazard 1 GHS Signal Word: Warning