Please login to your online account to display your discounted pricing

1-benzyl-3,5-dibromo-1H-1,2,4-triazole, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  MO07858DA

 View more versions of this product

Catalog No. MO07858DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 316.98
Quantity 1g
Assay 95%
CAS 106724-85-0
Assay Percent Range 95%
Chemical Name or Material 1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Molecular Formula C9H7Br2N3
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material 1-Benzyl-3,5-dibromo-1H-1,2,4-triazole
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H7Br2N3
Formula Weight 316.98
Accessories

GHS Signal Word: Warning

SDS & Safety Documents