missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-2-pyrrolidinone, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC154125000

 View more versions of this product

Catalog No. AC154125000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Benzyl-2-pyrrolidinone
Authentic
1.5500 to 1.5530 (20°C, 589nm)
500g
96.0
97%
MFCD00003195
LVUQCTGSDJLWCE-UHFFFAOYSA-N
1-benzylpyrrolidin-2-one
79176
Liquid
1.09g/mL
Glass bottle
Colorless to Yellow
5291-77-0
100.0
C11H13NO
1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
C1CC(=O)N(C1)CC2=CC=CC=C2
175.231
175.23
≥96.0%

Chemical Identifiers

5291-77-0
175.231
LVUQCTGSDJLWCE-UHFFFAOYSA-N
79176
C1CC(=O)N(C1)CC2=CC=CC=C2
C11H13NO
MFCD00003195
1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
1-benzylpyrrolidin-2-one
SDS Documents