Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-2-pyrrolidinone, 97%

$79.86 - $263.69

Chemical Identifiers

Chemical Name or Material 1-Benzyl-2-pyrrolidinone
CAS 5291-77-0
Molecular Formula C11H13NO
Formula Weight 175.23
MDL Number MFCD00003195
Beilstein 132168
Synonym N-Benzyl-gamma-butyrolactam; N-Benzyl-2-pyrrolidone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2353814

  SDS
alfa aesar™
B2353814*
25g Each for $79.86

AAB2353822

  SDS
alfa aesar™
B2353822
100g Each for $263.69
Description & Specifications

Specifications

Formula Weight 175.23
Health Hazard 1 Warning
Quantity 25g
Boiling Point 76° to 78°C (0.1mmHg)
Synonym N-Benzyl-gamma-butyrolactam; N-Benzyl-2-pyrrolidone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Benzyl-2-pyrrolidinone
Flash Point >110°C (230°F)
Molecular Formula C11H13NO
Beilstein 132168
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 5291-77-0
MDL Number MFCD00003195
Assay 97%

Applications
1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.