Please login to your online account to display your discounted pricing

1-Benzyl-1H-indol-5-ylamine, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO00835EA

 View more versions of this product

Catalog No. MO00835EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 222.29
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C15H14N2
Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
CAS 26807-73-8
MDL Number MFCD03070173
Packaging Amber Glass Bottle
Assay Percent Range 97%

Specifications

Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
CAS 26807-73-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H14N2
Formula Weight 222.29
MDL Number MFCD03070173