Please login to your online account to display your discounted pricing

1-Benzyl-1H-indol-5-ylamine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
CAS 26807-73-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C15H14N2
Formula Weight 222.29
MDL Number MFCD03070173
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00835EA

  SDS
fisher scientific company
MO00835EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.29
Quantity 10g
Assay 97%
CAS Min % 97.0
Molecular Formula C15H14N2
Chemical Name or Material 1-Benzyl-1H-indol-5-ylamine
CAS 26807-73-8
MDL Number MFCD03070173
Packaging Amber Glass Bottle
Assay Percent Range 97%