missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzyl-1H-indazol-3-ol, TCI America™

$41.25 - $113.06

Chemical Identifiers

CAS 2215-63-6
Molecular Formula C14H12N2O
Molecular Weight (g/mol) 224.263
MDL Number MFCD01631173
InChI Key SXPJFDSMKWLOAB-UHFFFAOYSA-N
Synonym 1-benzyl-3-hydroxy-1h-indazole, 1-benzyl-1h-indazol-3-ol, 1-benzyl-1,2-dihydro-3h-indazol-3-one, 1-benzylindazol-3-ol, 1-benzyl-1,2-dihydro-indazol-3-one, 1-benzyl-2,3-dihydroindazol-3-one, 1-benzyl-3-hydroxyindazole, chembl8612, 3h-indazol-3-one, 1,2-dihydro-1-phenylmethyl, benzyl-3-hydroxy-1h-indazole
PubChem CID 75181
IUPAC Name 1-benzyl-2H-indazol-3-one
SMILES C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=O)N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B24115G View Documents TCI America
B24115G
5g Each for $41.25
 
B241125G View Documents TCI America
B241125G
25g Each for $113.06
 
Specifications

Chemical Identifiers

2215-63-6
224.263
SXPJFDSMKWLOAB-UHFFFAOYSA-N
75181
C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=O)N2
C14H12N2O
MFCD01631173
1-benzyl-3-hydroxy-1h-indazole, 1-benzyl-1h-indazol-3-ol, 1-benzyl-1,2-dihydro-3h-indazol-3-one, 1-benzylindazol-3-ol, 1-benzyl-1,2-dihydro-indazol-3-one, 1-benzyl-2,3-dihydroindazol-3-one, 1-benzyl-3-hydroxyindazole, chembl8612, 3h-indazol-3-one, 1,2-dihydro-1-phenylmethyl, benzyl-3-hydroxy-1h-indazole
1-benzyl-2H-indazol-3-one

Specifications

Undesignated
5g
C14H12N2O
1-benzyl-3-hydroxy-1h-indazole, 1-benzyl-1h-indazol-3-ol, 1-benzyl-1,2-dihydro-3h-indazol-3-one, 1-benzylindazol-3-ol, 1-benzyl-1,2-dihydro-indazol-3-one, 1-benzyl-2,3-dihydroindazol-3-one, 1-benzyl-3-hydroxyindazole, chembl8612, 3h-indazol-3-one, 1,2-dihydro-1-phenylmethyl, benzyl-3-hydroxy-1h-indazole
C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=O)N2
224.263
224.26
≥98.0% (GC)
167°C
2215-63-6
MFCD01631173
SXPJFDSMKWLOAB-UHFFFAOYSA-N
1-benzyl-2H-indazol-3-one
75181
Crystal-Powder at 20°C
1-Benzyl-1H-indazol-3-ol
Safety and Handling

Safety and Handling

TSCA : No