missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 98%

$55.49 - $178.01

Chemical Identifiers

CAS 73953-89-6
Molecular Formula C11H9N3O4
Formula Weight 247.21
MDL Number MFCD00043512
Beilstein 29417
Chemical Name or Material 1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0843403 SDS Alfa Aesar™
L0843403
1g Each for $55.49
AAL0843406 SDS Alfa Aesar™
L0843406
5g Each for $178.01
Specifications Description & Specifications

Chemical Identifiers

73953-89-6
247.21
29417
C11H9N3O4
MFCD00043512
1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid

Specifications

73953-89-6
247.21
29417
1g
98%
C11H9N3O4
MFCD00043512
178° to 182°C
1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XZ4580000