missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0843406
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Benzyl-1,2,3-triazole-4,5-dicarboxylic acid | |
178°C to 182°C | |
MFCD00043512 | |
29417 | |
IYYBUXUZYWGZGB-UHFFFAOYSA-N | |
1-benzyltriazole-4,5-dicarboxylic acid | |
427031 | |
98% |
73953-89-6 | |
C11H9N3O4 | |
5 g | |
1-benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid, v-triazole-4,5-dicarboxylic acid, 1-benzyl, 1h-1,2,3-triazole-4,5-dicarboxylic acid, 1-phenylmethyl, 4-26-00-00965 beilstein handbook reference, 1-benzyl1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1,2,3-triazol-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3 triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid # | |
C1=CC=C(C=C1)CN2C(=C(N=N2)C(=O)O)C(=O)O | |
247.21 | |
247.21 |
Chemical Identifiers
73953-89-6 | |
247.21 | |
IYYBUXUZYWGZGB-UHFFFAOYSA-N | |
427031 | |
C1=CC=C(C=C1)CN2C(=C(N=N2)C(=O)O)C(=O)O |
C11H9N3O4 | |
MFCD00043512 | |
1-benzyl-1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid, v-triazole-4,5-dicarboxylic acid, 1-benzyl, 1h-1,2,3-triazole-4,5-dicarboxylic acid, 1-phenylmethyl, 4-26-00-00965 beilstein handbook reference, 1-benzyl1,2,3-triazole-4,5-dicarboxylic acid, 1-benzyl-1,2,3-triazol-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3 triazole-4,5-dicarboxylic acid, 1-benzyl-1h-1,2,3-triazole-4,5-dicarboxylic acid # | |
1-benzyltriazole-4,5-dicarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : XZ4580000
TSCA : No
Recommended Storage : Ambient temperatures