Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylpiperidine, 98%

$47.61 - $146.46

Chemical Identifiers

Chemical Name or Material 1-Benzoylpiperidine
CAS 776-75-0
Molecular Formula C12H15NO
Formula Weight 189.26
MDL Number MFCD00023702
Beilstein 136611
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1237803

  SDS
alfa aesar™
L1237803
1g Each for $47.61

AAL1237806

  SDS
alfa aesar™
L1237806
5g Each for $146.46
Description & Specifications

Specifications

Formula Weight 189.26
CAS 776-75-0
Chemical Name or Material 1-Benzoylpiperidine
Assay 98%
Molecular Formula C12H15NO
MDL Number MFCD00023702
Quantity 1g
Boiling Point 180° to 184°C (20mmHg)
Beilstein 136611
RTECS Number TM4600000
Melting Point 48° to 50°C