missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylpiperidine, 98%

Manufacturer:  Alfa Aesar™ L1237806

 View more versions of this product

Catalog No. AAL1237806



Specifications

Specifications

1-Benzoylpiperidine
48°C to 50°C
776-75-0
MFCD00023702
1-benzoylpiperidine, n-benzoylpiperidine, benzoylpiperidine, piperidine, 1-benzoyl, protectine i, phenyl piperidin-1-yl methanone, n-benzoylpiperidin, benzoic acid n-piperidide, alpha-repellin, benzoic acid, piperidide
C1CCN(CC1)C(=O)C2=CC=CC=C2
189.258
189.26
180°C to 184°C (20mmHg)
5g
C12H15NO
136611
YXTROGRGRSPWKL-UHFFFAOYSA-N
phenyl(piperidin-1-yl)methanone
69892
98%

Chemical Identifiers

776-75-0
189.258
YXTROGRGRSPWKL-UHFFFAOYSA-N
69892
C1CCN(CC1)C(=O)C2=CC=CC=C2
C12H15NO
MFCD00023702
1-benzoylpiperidine, n-benzoylpiperidine, benzoylpiperidine, piperidine, 1-benzoyl, protectine i, phenyl piperidin-1-yl methanone, n-benzoylpiperidin, benzoic acid n-piperidide, alpha-repellin, benzoic acid, piperidide
phenyl(piperidin-1-yl)methanone
Safety and Handling

Safety and Handling

RTECSNumber : TM4600000

Documents