missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzoylpiperazine +98%, ACROS Organics™

Chemical Identifiers

CAS 13754-38-6
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C11H14N2O
Formula Weight 190.25
MDL Number MFCD00810192
Chemical Name or Material 1-Benzoylpiperazine
Assay ≥98.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC384720050 SDS Thermo Fisher Scientific
AC384720050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

13754-38-6
100.0
C11H14N2O
MFCD00810192
≥98.0%
98.0
98+%
190.25
1-Benzoylpiperazine

Specifications

13754-38-6
100.0
98+%
190.25
Authentic
Yellow
5g
≥98.0%
98.0
Crystalline Powder
C11H14N2O
MFCD00810192
Glass bottle
62° to 66°C
1-Benzoylpiperazine

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger