missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylpiperazine, 97%

$41.31 - $564.12

Chemical Identifiers

CAS 13754-38-6
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
MDL Number MFCD00810192
InChI Key VUNXBQRNMNVUMV-UHFFFAOYSA-N
Synonym 1-benzoyl piperazine
PubChem CID 762654
IUPAC Name phenyl(piperazin-1-yl)methanone
SMILES C1CN(CCN1)C(=O)C2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5589603 View Documents Alfa Aesar™
H5589603
1g Each for $41.31
 
AAH5589606 View Documents Alfa Aesar™
H5589606
5g Each for $156.55
 
AAH5589614 View Documents Alfa Aesar™
H5589614
25g Each for $564.12
 
Specifications

Chemical Identifiers

13754-38-6
190.246
VUNXBQRNMNVUMV-UHFFFAOYSA-N
762654
C1CN(CCN1)C(=O)C2=CC=CC=C2
C11H14N2O
MFCD00810192
1-benzoyl piperazine
phenyl(piperazin-1-yl)methanone

Specifications

145°C (0.05mmHg)
1g
C11H14N2O
1-benzoyl piperazine
C1CN(CCN1)C(=O)C2=CC=CC=C2
190.246
190.24
1-Benzoylpiperazine
61°C to 64°C
13754-38-6
MFCD00810192
VUNXBQRNMNVUMV-UHFFFAOYSA-N
phenyl(piperazin-1-yl)methanone
762654
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures