missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylpiperazine, 97%

Manufacturer:  Alfa Aesar™ H5589603

 View more versions of this product

Catalog No. AAH5589603



Specifications

Specifications

1-Benzoylpiperazine
61°C to 64°C
13754-38-6
MFCD00810192
VUNXBQRNMNVUMV-UHFFFAOYSA-N
phenyl(piperazin-1-yl)methanone
762654
97%
145°C (0.05mmHg)
1g
C11H14N2O
1-benzoyl piperazine
C1CN(CCN1)C(=O)C2=CC=CC=C2
190.246
190.24

Chemical Identifiers

13754-38-6
190.246
VUNXBQRNMNVUMV-UHFFFAOYSA-N
762654
C1CN(CCN1)C(=O)C2=CC=CC=C2
C11H14N2O
MFCD00810192
1-benzoyl piperazine
phenyl(piperazin-1-yl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

Documents