Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylnaphthalene, 97%

$209.45 - $804.61

Chemical Identifiers

Chemical Name or Material 1-Benzoylnaphthalene
CAS 642-29-5
Molecular Formula C17H12O
Formula Weight 232.28
MDL Number MFCD00046431
Synonym 1-Naphthyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2212006

  SDS
alfa aesar™
B2212006
5g Each for $209.45

AAB2212014

  SDS
alfa aesar™
B2212014
25g Each for $804.61
Description & Specifications

Specifications

Formula Weight 232.28
Melting Point 75° to 76°C
Molecular Formula C17H12O
CAS 642-29-5
Chemical Name or Material 1-Benzoylnaphthalene
Health Hazard 1 Warning
Assay 97%
Synonym 1-Naphthyl phenyl ketone
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
MDL Number MFCD00046431
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.