missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoylimidazole, 96%

$51.97 - $51.97

Chemical Identifiers

CAS 10364-94-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD00014499
InChI Key JEGIFBGJZPYMJS-UHFFFAOYSA-N
Synonym n-benzoylimidazole, 1-benzoylimidazole, 1-benzoyl-1h-imidazole, 1h-imidazol-1-yl phenyl methanone, 1h-imidazole, 1-benzoyl, imidazol-1-yl phenyl methanone, 1h-imidazol-1-ylphenyl-methanone, imidazolyl phenyl ketone, pubchem7593, acmc-1bpvi
PubChem CID 66319
IUPAC Name imidazol-1-yl(phenyl)methanone
SMILES C1=CC=C(C=C1)C(=O)N2C=CN=C2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0065706 View Documents Alfa Aesar™
L0065706
5g Each for $51.97
 
Specifications

Chemical Identifiers

10364-94-0
172.187
JEGIFBGJZPYMJS-UHFFFAOYSA-N
66319
C1=CC=C(C=C1)C(=O)N2C=CN=C2
C10H8N2O
MFCD00014499
n-benzoylimidazole, 1-benzoylimidazole, 1-benzoyl-1h-imidazole, 1h-imidazol-1-yl phenyl methanone, 1h-imidazole, 1-benzoyl, imidazol-1-yl phenyl methanone, 1h-imidazol-1-ylphenyl-methanone, imidazolyl phenyl ketone, pubchem7593, acmc-1bpvi
imidazol-1-yl(phenyl)methanone

Specifications

Odorless
5g
C10H8N2O
125881
JEGIFBGJZPYMJS-UHFFFAOYSA-N
imidazol-1-yl(phenyl)methanone
66319
96%
120°C to 121°C (0.8mmHg)
10364-94-0
MFCD00014499
n-benzoylimidazole, 1-benzoylimidazole, 1-benzoyl-1h-imidazole, 1h-imidazol-1-yl phenyl methanone, 1h-imidazole, 1-benzoyl, imidazol-1-yl phenyl methanone, 1h-imidazol-1-ylphenyl-methanone, imidazolyl phenyl ketone, pubchem7593, acmc-1bpvi
C1=CC=C(C=C1)C(=O)N2C=CN=C2
172.187
172.19
1-Benzoylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : 2° to 8°C