missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Benzoyl-4-piperidone, TCI America™

$116.11 - $401.81

Chemical Identifiers

CAS 24686-78-0
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00006189
InChI Key NZAXGZYPZGEVBD-UHFFFAOYSA-N
Synonym n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
PubChem CID 90583
IUPAC Name 1-benzoylpiperidin-4-one
SMILES C1CN(CCC1=O)C(=O)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B37785G View Documents TCI America
B37785G
5g Each for $116.11
 
B377825G View Documents TCI America
B377825G
25g Each for $401.81
 
Specifications

Chemical Identifiers

24686-78-0
203.241
NZAXGZYPZGEVBD-UHFFFAOYSA-N
90583
C1CN(CCC1=O)C(=O)C2=CC=CC=C2
C12H13NO2
MFCD00006189
n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
1-benzoylpiperidin-4-one

Specifications

160°C
58°C
24686-78-0
MFCD00006189
NZAXGZYPZGEVBD-UHFFFAOYSA-N
1-benzoylpiperidin-4-one
90583
Crystal-Powder at 20°C
1-Benzoyl-4-piperidone
Yellow
5g
C12H13NO2
n-benzoyl-4-piperidone, 1-benzoyl-4-piperidinone, 4-piperidinone, 1-benzoyl, 1-benzoyl-4-piperidone, 1-benzoyl-4-oxopiperidine, 4-piperidone, 1-benzoyl, 1-phenylcarbonyl piperidin-4-one, 1-benzoyl-piperidin-4-one, n-benzoyl-4piperidone, 1-benzoyl-piperidinoine
C1CN(CCC1=O)C(=O)C2=CC=CC=C2
203.241
203.24
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No