Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Benzoyl-4-piperidone, 98+%

$122.51 - $1,237.58

Chemical Identifiers

Chemical Name or Material 1-Benzoyl-4-piperidone
CAS 24686-78-0
Molecular Formula C12H13NO2
Formula Weight 203.24
MDL Number MFCD00006189
Beilstein 158848
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1390106

 
alfa aesar™
A1390106
5g Each for $122.51

AAA1390114

 
alfa aesar™
A1390114
25g Each for $374.04

AAA1390122

 
alfa aesar™
A1390122
100g Each for $1,237.58
Description & Specifications

Specifications

Formula Weight 203.24
Chemical Name or Material 1-Benzoyl-4-piperidone
Assay ≥98%
Molecular Formula C12H13NO2
MDL Number MFCD00006189
CAS 24686-78-0
Beilstein 158848
Boiling Point 158° to 160°C (0.2mmHg)
Melting Point 52° to 57°C
Quantity 5g
Flash Point >110°C (230°F)