Please login to your online account to display your discounted pricing

1-Benzothiophene-5-carbaldehyde, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific uk ltd  CC24804DA

 View more versions of this product

Catalog No. CC24804DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 162.21
Quantity 1g
Assay 95%
Assay Percent Range 95%
Packaging Amber Glass Bottle
CAS 10133-30-9
Health Hazard 1 Warning
Chemical Name or Material 1-Benzothiophene-5-carbaldehyde
Molecular Formula C9H6OS

Specifications

Chemical Name or Material 1-Benzothiophene-5-carbaldehyde
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H6OS
Formula Weight 162.21
Accessories

Warning

SDS & Safety Documents