missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Benzothiophene-3-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12301EA
Specifications
1-Benzothiophene-3-carboxylic acid | |
96.5 | |
97% | |
C9H6O2S | |
10g | |
DRBLTQNCQJXSNU-UHFFFAOYSA-N | |
1-benzothiophene-3-carboxylic acid | |
601280 | |
97% |
5381-25-9 | |
100.0 | |
Amber Glass Bottle | |
MFCD01846406 | |
benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid | |
C1=CC=C2C(=C1)C(=CS2)C(=O)O | |
178.205 | |
178.21 |
Chemical Identifiers
5381-25-9 | |
178.205 | |
DRBLTQNCQJXSNU-UHFFFAOYSA-N | |
601280 | |
C1=CC=C2C(=C1)C(=CS2)C(=O)O |
C9H6O2S | |
MFCD01846406 | |
benzo b thiophene-3-carboxylic acid, benzothiophene-3-carboxylic acid, benzothiophene-3-carboxylicacid, pubchem13463, acmc-20a10z, ksc178m8d, 1-benzo b thiophene-3-carboxylic acid, #, thianaphthene-3-carboxylic acid | |
1-benzothiophene-3-carboxylic acid |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously
Warning