Please login to your online account to display your discounted pricing

1-Benzothiophene-3-carbonyl chloride, Maybridge


Quantity 1g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CC12302DA

 View more versions of this product

Catalog No. CC12302DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 196.66
Quantity 1g
Assay 95%
CAS Max % 100.0
Chemical Name or Material 1-Benzothiophene-3-carbonyl chloride
Assay Percent Range 95%
Molecular Formula C9H5ClOS
MDL Number MFCD02682005
CAS 39827-12-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Packaging Amber glass bottle
CAS Min % 94.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Benzothiophene-3-carbonyl chloride
CAS 39827-12-8
CAS Min % 94.5
CAS Max % 100.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C9H5ClOS
Formula Weight 196.66
MDL Number MFCD02682005
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.